Key Documents
SYNF964014
SynthwareBuchner funnel with perforated plate
60 mL, joint: ST/NS 14/20, plate O.D. 42 mm
Synonyme(s) :
Synthware F964014, filter funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F964014
Capacité
60 mL
60 mL
D.E. de la plaque/du plateau
42 mm
Description générale
Synthware Büchner funnel with an inner joint and a perforated glass plate, sealed with a perforated glass disc to support filter media. It features a standard taper inner joint (14/20) and a hose connector for the vacuum source.
Informations légales
Synthware is a trademark of Kemtech America Inc.