Key Documents
SYNF932490
Synthware filter funnel with perforated plate
joint: ST/NS 24/40, plate O.D. 90 mm
Synonyme(s) :
Synthware F932490
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F932490
Diam. du disque
24 mm
D.E. de la plaque/du plateau
90 mm
Description générale
O.D. of disc is 24 mm.
Informations légales
Synthware is a trademark of Kemtech America Inc.