SYNF701420M
Synthware™ fritted filter tube
joint: ST/NS 14/20, disc O.D. 20 mm, porosity: medium
Synonyme(s) :
Synthware F701420M, straight filter tube
About This Item
Produits recommandés
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F701420M
D.E. du disque
20 mm