Principaux documents
SYNF672460
Synthware pressure equalizing addition funnel with PTFE stopcock in equalizing line
60 mL, joint: ST/NS 24/40, stopcock: PTFE
Synonyme(s) :
Synthware F672460, addition funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F672460
Capacité
60 mL
60 mL
Description générale
With straight bore PTFE stopcock in pressure equalizing line, graduated.
Informations légales
Synthware is a trademark of Kemtech America Inc.