Principaux documents
SYNF490500
Synthware storage flask with high vacuum valve and side arm
500 mL, valve size 0-4 mm
Synonyme(s) :
Synthware F490500, round bottom flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
round bottom
single-neck (high vacuum valve)
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F490500
Capacité
500 mL
500 mL
Taille de la vanne
0-4 mm
Description générale
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper( “umbrella”) to protect Viton® O-ring on shaft..
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company