Principaux documents
SYNF311925
Synthware recovery flask
25 mL, joint: ST/NS 19/22
Synonyme(s) :
Synthware F311925, oval shape flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F311925
Capacité
25 mL
25 mL
Catégories apparentées
Description générale
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.