Key Documents
SYNF301025
Synthware single neck round bottom flask
capacity 25 mL, joint: ST/NS 14/10
Synonyme(s) :
Synthware F301025, glass round flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS14/10
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F301025
Capacité
25 mL
Description générale
Supplied with one holed compression caps, one Viton® O-rings and one PTFE - faced silicone rubber septum. For replacement caps, see RC112; for Viton® O-rings, see R014; for septa, see R1420.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company