Key Documents
SYNF122450
Synthware single-neck flat-bottom flask
capacity 50 mL, joint: ST/NS 24/40
Synonyme(s) :
Synthware F122450, glass round flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
flat bottom
single-neck
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F122450
Capacité
50 mL
Description générale
Heavy wall single neck, flat bottom flask with an outer joint. Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.