Principaux documents
SYNC154200
Synthware West condenser
joint: ST/NS 24/40, jacket L 200 mm
Synonyme(s) :
Synthware C154200, distillation condenser, Inland Revenue condenser
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C154200
L de l'enveloppe
200 mm
Catégories apparentées
Description générale
With narrow annular space for fast coolant speed, thus high cooling efficiency.
Informations légales
Synthware is a trademark of Kemtech America Inc.