SYNB310350C
Synthware™ gas washing bottle with fritted disc
joint: ST/NS 24/40, capacity 350 mL, Disc porosity C
Synonyme(s) :
Synthware B310350C, Drechsel bottle, gas washing bottle
About This Item
Produits recommandés
Matériaux
clear bottle
cylindrical bottle
straight bottle
Joint
joint: ST/NS24/40
Caractéristiques
cap
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Synthware B310350C
Hauteur
250 mm
Capacité
350 mL
Catégories apparentées
Description générale
Synthware™ Gas washing bottle with an extra-large diameter (60mm) fritted disc sealed into the bottom of the bottle to enable increased gas throughput and uniform gas distribution. The 24/40 standard taper top outer joint accepts a A292440 gas inlet adapter which is supplied with bottle.
Informations légales
Synthware is a trademark of Kemtech America Inc.