Principaux documents
SYNA282250
Synthware anti-splash adapter
250 mL, top joint: ST/NS 29/42, bottom joint: ST/NS 24/40
Synonyme(s) :
Synthware A282250, rotary evaporator bump trap, bump trap
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS24/40
joint: ST/NS top29/42
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A282250
Capacité
250 mL
Catégories apparentées
Description générale
This design prevents possible contamination from rotary evaporator.
Informations légales
Synthware is a trademark of Kemtech America Inc.