Principaux documents
DWK73660-18
KIMBLE® KIM-KAP® push cap
assorted colors polypropylene cap, cap diam. (18 mm)
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
assorted colors polypropylene cap
Stérilité
non-sterile
Conditionnement
case of 1000 ea
Fabricant/nom de marque
Kimble® 73660-18
Diam. du bouchon
(18 mm)
Description générale
  • KIM-KAP polypropylene closures for culture tubes
  • Autoclavable
  • Color coded
Informations légales
KIM-KAP is a registered trademark of DWK LIFE SCIENCES LLC
KIMBLE is a registered trademark of DWK Life Sciences