Principaux documents
DWK2518415-B
Kimble® Closed Top Linerless Polypropylene Screw Thread Cap
18-415, black screw cap
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.17
Matériaux
black screw cap
red polypropylene screw cap (Push fit)
Conditionnement
pkg of 1 packs
Fabricant/nom de marque
Kimble® 2518415-B
Raccords
thread
Application
Closure solution for vials and bottles
Caractéristiques et avantages
  • Economical, one-piece construction
  • Unique design provides exceptional sealing properties
  • Autoclavable
  • Designed from ASTM Specification E982
Informations légales
KIMBLE is a registered trademark of DWK Life Sciences