Key Documents
DWK213544106
DURAN® powder funnel with short wide stem
glass funnel(s), funnel capacity (75 mL)
Synonyme(s) :
213544106
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Matériaux
glass funnel(s)
Conditionnement
case of 10 ea
Fabricant/nom de marque
DURAN® 213544106
Capacité de l'entonnoir
(75 mL)
Informations légales
DURAN is a registered trademark of DWK Life Sciences