Key Documents
BR708212
Filling tube for Dispensette® S Organic / S Organic
1, 2, 5 and 10 mL, FEP tube, L 125-240 mm, individual adjustment of length
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Matériaux
FEP tube
Fabricant/nom de marque
BRAND 708212
Conditionnement
1 ea of
Longueur
125-240 mm
D.E.
6 mm
Capacité
1, 2, 5 and 10 mL
Description générale
Adjusts to various bottle heights.
Informations légales
Dispensette is a registered trademark of BRAND GMBH + CO KG