BR474410
BRAND® plastic staining trough, Schiefferdecker pattern
with lid
Synonyme(s) :
Microscopy, Staining, Staining Accessories
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
MA.01
Produits recommandés
Matériaux
PMP
Caractéristiques
with lid
Fabricant/nom de marque
BRAND 474410
L × l
76 mm × 26 mm , holds 20 slides
Catégories apparentées
Description générale
The BRAND® Plastic Staining Trough in the Schiefferdecker pattern is designed for efficient staining of up to 20 slides, organized in 10 pairs back-to-back. With dimensions of 76 x 26 mm, this trough provides ample space for optimal staining while allowing easy access to each slide. Its robust construction ensures durability and reliability in laboratory settings, making it an essential tool for histology and microscopy applications.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG