Key Documents
BR114866
BRAND® storage box
Holds 100 x 1.2-2 mL tubes
Synonyme(s) :
Accessories, Cryogenic Tubes, Sample Storage, Storing box, Tubes
About This Item
Code UNSPSC :
41123300
Nomenclature NACRES :
NB.51
Matériaux
polycarbonate
Caractéristiques
Holds 100 x 1.2-2 mL tubes
Conditionnement
pack of 4 ea
Fabricant/nom de marque
BRAND 114866
Taille
132x132x52 mm
Description générale
For cryogenic tubes.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG