Merck
  • Neurons under T Cell Attack Coordinate Phagocyte-Mediated Synaptic Stripping.
Neurons under T Cell Attack Coordinate Phagocyte-Mediated Synaptic Stripping.
Cell (2018-09-04)
Giovanni Di Liberto, Stanislav Pantelyushin, Mario Kreutzfeldt, Nicolas Page, Stefano Musardo, Roland Coras, Karin Steinbach, Ilena Vincenti, Bogna Klimek, Thomas Lingner, Gabriela Salinas, Nathalie Lin-Marq, Ori Staszewski, Marta Joana Costa Jordão, Ingrid Wagner, Kristof Egervari, Matthias Mack, Camilla Bellone, Ingmar Blümcke, Marco Prinz, Daniel D Pinschewer, Doron Merkler
ABSTRACT
Inflammatory disorders of the CNS are frequently accompanied by synaptic loss, which is thought to involve phagocytic microglia and complement components. However, the mechanisms accounting for aberrant synaptic connectivity in the context of CD8+ T cell-driven neuronal damage are poorly understood. Here, we profiled the neuronal translatome in a murine model of encephalitis caused by CD8+ T cells targeting antigenic neurons. Neuronal STAT1 signaling and downstream CCL2 expression were essential for apposition of phagocytes, ensuing synaptic loss and neurological disease. Analogous observations were made in the brains of Rasmussen's encephalitis patients. In this devastating CD8+ T cell-driven autoimmune disease, neuronal STAT1 phosphorylation and CCL2 expression co-clustered with infiltrating CD8+ T cells as well as phagocytes. Taken together, our findings uncover an active role of neurons in coordinating phagocyte-mediated synaptic loss and highlight neuronal STAT1 and CCL2 as critical steps in this process that are amenable to pharmacological interventions.
Product Description
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-P2RY12 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution