Skip to Content
Merck
All Photos(1)

Key Documents

818207

TSKgel® HPLC Columns Hardware and Accessories

pkg of 3 ea

Synonym(s):

Guardfilter for 4.6 mm I.D. TSKgel® Super Series columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

TSKgel® Hardware and Accessories, pkg of 3 ea

material

stainless steel

product line

TSKgel®

packaging

pkg of 3 ea

manufacturer/tradename

Tosoh 18207

technique(s)

HPLC: suitable

pore size

140 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Use to protect TSKgel Super-ODS, Super-Octyl and Super-Phenyl columns. Requires Guardfilter Holder PN 818206; sold separately.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

required but not provided

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service