Skip to Content
Merck
All Photos(1)

Key Documents

22052

Swagelok® Zero-Dead Volume Union

Swagelok® I Of-6-GC, stainless steel, 1/16 in.

Synonym(s):

Swagelok® 100-6,SS 0-DEAD VOL UNION 1/16" EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717
eCl@ss:
32035302

material

stainless steel

description

zero dead volume

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® I Of-6-GC

fitting

1/16 in.

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Swagelok® zero-dead- volume unions are principally designed for liquid and gas chromatography. On application of Swagelok® zero-dead- volume unions, a perfect low-dead-volume connection between column terminator and injector or detector is obtained.

Application

Swagelok® zero-dead- volume unions maybe used in all the connections between the valve, the columns and the detector during the studies pertaining to the relation between column plate number, particle size, and loadability for a given solute.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Instrumentation for High Performance Liquid Chromatography.
Huber JFK
Science, 87-88 (2011)
Column loadability and particle size in preparative liquid chromatography.
De-Jong AW
Journal of Chromatography A, 209 (3), 432-436 (1981)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service