Key Documents
TMO140080
Nunc® EasyFill Cell Factory filter assembly cap
high-density polyethylene, pore size 0.22 μm
Synonym(s):
closed cell system cap, closed cell system filter cap
About This Item
UNSPSC Code:
41161502
NACRES:
NB.13
Pricing and availability is not currently available.
material
high-density polyethylene
packaging
pack of 2 ea
manufacturer/tradename
Nunc 140080
pore size
0.22 μm pore size
General description
Further the development of applications that require a more closed cell culture system using Thermo Scientific Nunc Cell Factory Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)