Key Documents
EP5427717005
Eppendorf® Adapter
Holds PCR Strips tubes, pack of 4 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
Holds PCR Strips tubes
packaging
pack of 4 ea
manufacturer/tradename
Eppendorf® 5427717005
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with Rotor F-45-64-5-PCR
Legal Information
Eppendorf is a registered trademark of Eppendorf AG