EP5409706004
Rotor for Eppendorf® 5427R Centrifuge
FA-45-30-11, aerosol tight, to hold, 30 x 1.5/2.0 mL tubes, lid
All Photos(1)
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Recommended Products
feature
lid
to hold 30 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5409706004 (Global)
parameter
14,000 rpm
20,800 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5427R
General description
Rotors to fit Eppendorf 5427 R refrigerated centrifuge.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)