Skip to Content
Merck
All Photos(1)

Key Documents

CLS3768

Corning® PDL 384 well plates

black polystyrene, flat bottom, sterile, lid, case of 100, pack of 20

Synonym(s):

384 multiwell plates, 384 well immunoassay plates, 384 well microplates, 384 well microtiter plates, 384 well plates, immunoassay plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.15
material:
black polystyrene
flat bottom
size:
384 wells

material

black polystyrene
flat bottom

sterility

sterile

feature

lid
plate format: low flange, with lid

packaging

case of 100
pack of 20

manufacturer/tradename

Corning 3768

size

384 wells

color

black

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service