Skip to Content
Merck
All Photos(1)

Key Documents

KARPAS 417

6100302, human blood, Single round to polymorphic cells growing in suspension

Synonym(s):

KARPAS, 417

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

KARPAS 417, 06100302, from human blood

biological source

human blood

growth mode

Suspension

karyotype

Complex translocations, refer to Karpas et al., 2005

morphology

Single round to polymorphic cells growing in suspension

products

Secretes IgGks

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

Human myeloma

Cell Line Description

Established from the peripheral blood of a 56 year-old male diagnosed with multiple myeloma.

DNA Profile

Not specified

Culture Medium

RPMI 1640 + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 0.5 - 2 x 1000,000 cells/ml; 5% CO2; 37 °C. Doubling time approximately 60 hours. Split saturated culture 1:2 every 2-4 days. Round to polymorhpic cells growing singly or in small clusters in suspension.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service