U-BLC1
6013102, human bladder, Epithelial
Synonym(s):
Human U-BLC1, U-BLC1 Cell Line
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Product Name
U-BLC1, 06013102
biological source
human bladder
growth mode
Not specified
karyotype
Aneuploid, with specific marker chromosomes
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human bladder transitional cell carcinoma
Cell Line Description
Spontaneously immortal cell line established from a primary transitional cell carcinoma of the bladder. Derived from a female patient.
Culture Medium
2/3 DMEM:1/3 RPMI + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. The medium must be changed to fresh medium every 48 hours.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service