Skip to Content
Merck
All Photos(1)

Key Documents

BF-2

21713, fish caudal trunk, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

BF-2, 00021713

biological source

fish caudal trunk

packaging

tube of 5 μg 00021713-DNA-5UG
pkg of vial of cells 00021713-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Fish bluegill fry

Cell Line Description

The cell line BF-2 has been derived from a trypsinised suspension of pooled caudal portions of the trunk of 1 year old fingerlings (Bluegill fry, Lepomis macrochirus). Cells have been reported to be susceptible to a variety of fish viruses including viral haemorrhagic septicaemia virus (VHSV), infectious hematopoietic necrosis virus (IHNV), spring viraemia of carp virus (SVCV) and pike fry rhabdovirus (PFRV).

Application

Fish virus studies

Culture Medium

EMEM(HBSS) (M5775) or EMEM (EBSS) (M2279) + 2mM L-Glutamine (G7513) + 850 mg/L Sodium Bicarbonate(NaHCO3) (S8761) + 1% Non Essential Amino Acids(NEAA) (M7145) + 120 mg/l Sodium Pyruvate(NaP) (S8636) + 10% FBS / FCS (F2442); a mixture of MEM (HBSS) and MEM (EBSS) with 1-2% FBS is best for stability of monolayer and maintenance medium

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:6 using 0.25% trypsin or trypsin/EDTA; cells replicate at 15 - 33°C, optimal temperature is 23 - 26°C; monolayer is stable for at least 10 days in maintenance medium (at 15 or 16°C for about 21 days). Fish c

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service