AXYSCT200SSA
Screw cap tubes and caps with O-rings, self-standing
size 2.0 mL, assorted colors, pkg of 8x500caps/cs
Synonym(s):
centrifuge tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
Axygen Inc SCT-200-SS-A
Corning
packaging
pkg of 8x500caps/cs
size
2.0 mL
color
assorted colors
Looking for similar products? Visit Product Comparison Guide
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service