Skip to Content
Merck
All Photos(1)

Key Documents

NIST3246

Ginkgo biloba (leaves)

NIST® SRM® 3246

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 3 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for use in validating analytical methods for the determination of flavonoids, terpene lactones, and toxic elements in Ginkgo biloba and similar matrices. This SRM can be used for quality assurance when assigning values to in-house control materials. This SRM has also been characterized for its DNA sequence. A unit of SRM 3246 consists of five bottles, each containing approximately 3 g of ground leaves. For more information, please refer to the SDS and COA.

SRM 3246_cert

SRM 3246_SDS

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: Bilobalide, Ginkgolide A, Ginkgolide B, Ginkgolide C, Ginkgolide J, Isorhamnetin, Kaempferol, Quercetin, Total Aglycones, Total Terpene Lactones
Trace Elements: Cadmium (Cd), Lead (Pb), Mercury (Hg)

Matrix Group: Botanicals

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service