XX1012503
Millipore Spring Clamp for Vacuum Filtration
25 mm, Aluminum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24
Recommended Products
material
anodized aluminum
technique(s)
vacuum filtration: suitable
diam.
25 mm
application(s)
filtration
shipped in
ambient
General description
Millipore Glass Vacuum Filtration Accessory Product
Spring Clamp, 25 mm, Aluminum
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
This item is an accessory part for the Millipore 25 mm Glass Microanalysis Filter Holder Kits and Millipore 25 mm Analytical Stainless Steel Filter Holder Kit.
Spring Clamp, 25 mm, Aluminum
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
This item is an accessory part for the Millipore 25 mm Glass Microanalysis Filter Holder Kits and Millipore 25 mm Analytical Stainless Steel Filter Holder Kit.
Application
25 mm, Aluminum
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service