Skip to Content
Merck
All Photos(1)

Key Documents

MABT65

Sigma-Aldrich

Anti-CEACAM1 Antibody, clone 4D1/C2

clone 4D1/C2, from mouse

Synonym(s):

Carcinoembryonic antigen-related cell adhesion molecule 1, Biliary glycoprotein 1, BGP-1, CD66a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4D1/C2, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CEACAM1(634)

Related Categories

General description

CEACAM1 (carcinoembryonic antigen-related cell adhesion molecule 1) is thought to be a member of the immunoglobulin superfamily and is known as an epithelial tumor suppressor and an angiogenic growth factor. It has also been linked to the actin-based cytoskeleton. CEACAM1 is also known as a cellular receptor for a number of human mucosa pathogenic bacteria. The loss of activity of CEACAM1 has been related to the development of colorectal cancer.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human CEACAM1.

Application

Detect CEACAM1 using this Anti-CEACAM1 Antibody, clone 4D1/C2 validated for use in WB, IH.
Immunohistochemistry Analysis: A representative lot was used by an independent laboratory to detect CEACAM1 in non-small-cell lung cancer (Dango, S., et al. (2008). Lung Cancer. 60:426-433).

Western Blot Analysis: A representative lot was used by an independent laboratory to detect CEACAM1 in human granulocyte lysate (Bogoevska, V. et al., (2006). Glycobiology. 16(3): 197–209).

Flow Cytometry Analysis: A representative lot was used by an independent laboratory to detect CEACAM1 in human granulocytes by FACS analysis (Bogoevska, V. et al., (2006). Glycobiology. 16(3): 197–209).
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blot in HepG2 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected CEACAM1 in 10 µg of HepG2 cell lysate.

Target description

~150 kDa observed.
Bands are usually observed at ~120-160 kDa, resulting from different glycosylations (Lauke, H., et al. (2004). Mol. Hum. Reprod. 10(4):247-252).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria L Dupuis et al.
Journal of immunotherapy (Hagerstown, Md. : 1997), 38(9), 357-370 (2015-10-09)
Several lines of evidence show that de novo expression of carcinoembryonic antigen-related cell adhesion molecule 1 (CEACAM1) is strongly associated with reduced disease-free survival of patients affected by metastatic melanoma. Previously published investigations report that homophilic interactions between CEACAM1 expressed
Max Klapholz et al.
The Journal of pathology, 257(2), 186-197 (2022-02-05)
Colorectal carcinoma (CRC) is the second leading cause of cancer mortality worldwide. CRC is stratified into two major groups: microsatellite stable (MSS) and microsatellite instability-high (MSI-H). MSS CRC constitutes the majority of cases, has worse overall prognosis, and thus far

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service