Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB217F

Sigma-Aldrich

Milli-Mark® Anti-CD235a-FITC Antibody, clone JC159

clone JC159, Milli-Mark®, from mouse

Synonym(s):

Glycophorin-A, PAS-2, Sialoglycoprotein alpha, MN sialoglycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44
conjugate:
FITC conjugate
application:
FACS
clone:
JC159, monoclonal
species reactivity:
human
citations:
1
technique(s):
flow cytometry: suitable

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

clone

JC159, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Glycophorin A, CD235a, is a single-pass membrane sialoglycoprotein and carrier of the blood group specificities M and N. The gene is located on chromosome 4q28-q31 and share sequence homology with genes encoding glycophorin B and glycophorin E (van der Schoot, 2002). Glycophorin A is expressed by erythroid cells beginning on morphologically recognizable erythroid precursors, just after the CFU-E stage, to the mature erythrocyte. Once glycophorin A is maximally expressed, the amount in each erythroid cell remains constant and does not appear to change during further maturation (Loken, 1987).

Specificity

Antibody recognizes Human CD235a antigen.

Immunogen

Membrane extracts of hairy cell leukemia prepared from human spleen

Application

Detect CD235a using this Milli-Mark Anti-CD235a-FITC Antibody, clone JC159 validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human lymphocytes

Target description

16 kDa (calculated)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Human Lymphocytes

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asma S Al-Amoodi et al.
iScience, 27(2), 108882-108882 (2024-02-07)
For almost two decades, clinicians have overlooked the diagnostic potential of CD34neg hematopoietic stem cells because of their limited homing capacity relative to CD34posHSCs when injected intravenously. This has contributed to the lack of appeal of using umbilical cord blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service