Skip to Content
Merck
All Photos(1)

Key Documents

B04F-01-5PK

Millipore

CellASIC ONIX B04F Microfluidic Bacteria Plate (4 chamber)

The B04F plates utilize microfabricated PDMS pillars to trap bacteria (or similarly sized cells) within a single focal plane for real time imaging and analysis

Synonym(s):

Microfluidic Bacteria Plate, ONIX B04F Plate, ONIX Bacteria Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.22
Pricing and availability is not currently available.

material

PMMA
glass
polycarbonate
polydimethylsiloxane (PDMS)

manufacturer/tradename

CellASIC® ONIX

W × L × H

85.2  mm × 127.3 mm × 14.3  mm

compatibility

for use with cells sized up to 0.7–3.5 µm

General description

The CellASIC® ONIX B04F Microfluidic Plate is a 4-chamber cell culture plate designed for use with the CellASIC® ONIX Microfluidic System and CellASIC® ONIX F84 Manifolds for enabling perfusion-based, long-term, live-cell analysis with solution switching that monitors trapped cells growing into biofilms.

Application

This bio-inspired plate provides a controlled and dynamic microenvironment for cells. The easy-to-use format and superior technology redefine the standard for microfluidics based experimentation.

Applications

  • Time-lapse analysis of bacteria cells
  • Temperature and gas atmospheric control (temperature shift, anoxic conditions etc.)
  • Long-term continuous perfusion experiments
  • Solution exchange experiments (induction, inhibition, drug dosing, etc.)
  • Comparison of up to 4 different cell types or exposure conditions (media components) in parallel
  • Cell division tracking (follow cell division over generations)
Research Category
Cell Culture

Legal Information

CELLASIC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service