Skip to Content
Merck
All Photos(2)

Key Documents

AB9913

Sigma-Aldrich

Anti-TNRC6B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Trinucleotide repeat-containing gene 6B protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, ox

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TNRC6B(23112)

General description

TNRC6B (trinucleotide repeat containing 6B) is a argonaute-associated protein that localizes to cytoplasmic P bodies and is essential for control of miRNA-guided mRNA cleavage. TNRC6B is one of the mammalian homologs of the metazoan GW182 and is required for well-organized miRNA repression. Studies suggest that TNRC6B-mediated miRNA silencing is promoted by the C-terminal silencing domain.

Specificity

Antibody recognizes TNRC6B

Immunogen

Epitope: N-terminan domain of TNRC6B
KLH-conjugated linear peptide corresponding to human TNRC6B.

Application

Anti-TNRC6B Antibody is a Rabbit Polyclonal Antibody for detection of TNRC6B also known as Trinucleotide repeat-containing gene 6B protein & has been validated in WB, ICC & IP.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected TNRC6B in HeLa cells. (Zipprich, J., et al. (2009). RNA. 15:781–793.)
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected TNRC6B on 10 µg of HeLa cell lysate.

Target description

~150/190 kDa observed. (Meister, G., et al. (2005). Current Biology. 15:2149–2155). An uncharacterized band at ~39 kDa may be observed in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alberto Parras et al.
Nature, 560(7719), 441-446 (2018-08-17)
Common genetic contributions to autism spectrum disorder (ASD) reside in risk gene variants that individually have minimal effect sizes. As environmental factors that perturb neurodevelopment also underlie idiopathic ASD, it is crucial to identify altered regulators that can orchestrate multiple

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service