Skip to Content
Merck
All Photos(2)

Key Documents

8.00866

Sigma-Aldrich

Diisopropyl ether

(stabilized with 2,6-di-tert-butyl-4-methylphenol (BHT)) for synthesis

Synonym(s):

2,2′-Oxybis[propane], 2,2-Propoxypropane, Isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731256
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-560-6
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
68-69 °C (lit.)
vapor pressure:
120 mmHg ( 20 °C)

grade

for synthesis
synthesis grade

Quality Level

vapor density

3.5 (vs air)

vapor pressure

120 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

827 °F

potency

5880 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

expl. lim.

1-21 %, 100 °F

dilution

(for synthesis)

refractive index

n20/D 1.367 (lit.)

pH

7 (20 °C)

bp

68-69 °C (lit.)

mp

−85 °C (lit.)

transition temp

flash point -28 °C

solubility

3.11 g/L

density

0.725 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(C)OC(C)C

InChI

1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

General description

Diisopropylether (DIPE) is secondary ether. Besides its high-octane number, it has low toxicity, low production cost, and good anti-knock properties. It is flammable, non-reactive, colorless, and slightly soluble in water but has a high solubility in organic compounds. Due to its ability to liquefy oil-based solutions, it is being used as a solvent in paints, waxes, dyes, and stain removal. DIPE serves as a solvent in various chemical processes, also used as an extractant and oxygenate in motor fuel.

Application

Diisopropyl ether is used as a solvent in the aza-Michael addition of mono-and bifunctional amines to acrylate.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.722 - 0.724
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermophysical properties of ternary liquid mixture of diisopropyl ether (1)+ ethanol (2)+ n-heptane (3): Measurement and correlation
Kashyap P, et al.
Journal of Molecular Liquids, 319, 114219-114219 (2020)
Thermodynamic and acoustic properties of binary mixtures of diisopropyl ether, benzene and alkanes at 298.15, 308.15 and 318.15 K: Prigogine-Flory-Patterson theory and graph theory
Devi R, et al.
Journal of Molecular Liquids, 275, 364-377 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service