Skip to Content
Merck
All Photos(1)

Key Documents

IMF-020S

Supelco

iMethod Test Kit for Benzodiazepines with Synthetic Urine

kit of 23 × 1.0 mL ampules, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

packaging

kit of 23 × 1.0 mL ampules

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

General description

A kit of certified reference solutions suitable for use with the Benzodiazepine iMethod® application.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
iMethod is a registered trademark of CEM Corporation

Kit Components Also Available Separately

Product No.
Description
SDS

  • A-903Alprazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • A-907alpha-Hydroxyalprazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • A-9117-Aminoflunitrazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • A-9167-Aminoclonazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • C-907Clonazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • D-907Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • F-003Flurazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • F-907Flunitrazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • L-901Lorazepam solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • M-908Midazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • N-905Nordiazepam solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • O-902Oxazepam solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • T-907Temazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • T-911α-Hydroxytriazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • A-902Alprazolam-d5 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • A-904alpha-Hydroxyalprazolam-d5 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • A-9177-Aminoclonazepam-d4 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • A-9217-Aminoflunitrazepam-d7 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • D-902Diazepam-d5 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • F-915Flunitrazepam-d7, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • N-903Nordiazepam-d5 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • O-901Oxazepam-d5 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • Surine Negative Urine Control, bottle of 50 mL, Cerilliant®

See All (23)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service