Skip to Content
Merck
All Photos(1)

Key Documents

860663P

Avanti

Sphingosine (d22:1)

Avanti Research - A Croda Brand 860663P, powder

Synonym(s):

D-erythro-Sphingosine (C22 base)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H45NO2
CAS Number:
Molecular Weight:
355.60
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860663P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860663P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCCCCCCC

InChI

1S/C22H45NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-22(25)21(23)20-24/h18-19,21-22,24-25H,2-17,20,23H2,1H3/b19-18+/t21-,22+/m0/s1

InChI key

BSHKJMWKIAPLFB-ICWQGCNBSA-N

General description

Sphingosine (d22:1) is a sphingosine with 20 carbon long chain bases (LCB). It is a component of Asterias amurensis glucosylceramide.

Biochem/physiol Actions

Sphingosine negatively regulates cell proliferation and induces apoptosis. It has an ability to regulate the activities of phospholipases, protein kinases, ion channels, cannabinoid receptor type 1 (CB-1) receptors and steroidogenic factor 1 (SF-1) receptors.

Packaging

5 mL Amber Glass Screw Cap Vial (860663P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Mikami et al.
Journal of oleo science, 65(8), 671-680 (2016-07-20)
Asterias amurensis starfish provide several bioactive species in addition to being fishery waste. Glucosyl ceramides (GlcCers) were extracted from the viscera of these starfish and were isolated by silica gel column chromatography. Degraded GlcCers generated A. amurensis sphingoid bases (ASBs)
the
Lipid Mediators and Their Metabolism in the Brain (2011)
Olivier Cuvillier
Biochimica et biophysica acta, 1585(2-3), 153-162 (2003-01-18)
The sphingolipid metabolites ceramide, sphingosine, and sphingosine 1-phosphate contribute to controlling cell proliferation and apoptosis. Ceramide and its catabolite sphingosine act as negative regulators of cell proliferation and promote apoptosis. Conversely, sphingosine 1-phosphate, formed by phosphorylation of sphingosine by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service