Skip to Content
Merck
All Photos(1)

Key Documents

Z507776

Ace gas dispersion tube with porous fritted glass tip, 15° bend

fits flask, 3000 ‑ 5000 mL, porosity 145-174 μm (A)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

feature

fits flask 3000 ‑ 5000 mL

manufacturer/tradename

Ace Glass 720407

O.D. × L

7 mm × 330 mm

pore size

145-174 μm porosity (A)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Fit #7 Ace-Thred

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service