Key Documents
Z319449
Büchi® evaporating flask, pear-shaped
glass, volume 250 mL, joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
glass
pear-shaped
single-neck
Joint
joint: ST/NS29/32
packaging
pack of 1 ea
manufacturer/tradename
Büchi 00433
capacity
250 mL
volume
250 mL
General description
These flasks fit R200/205, R210/215, and RII evaporators
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)