WHA5230185
Whatman® filter paper, Grade 230, creped
circles, diam. 185 mm (7.3 in.), pkg of 50 ea
Synonym(s):
Whatman filter, Whatman paper, Z761958, paper filter
About This Item
Recommended Products
material
circles
creped white cellulose
feature
Grade 230
packaging
pkg of 50 ea
manufacturer/tradename
Whatman 5230-185
parameter
fast flow rate
fast flow rate
diam.
185 mm (7.3 in.)
thickness
0.40 mm (0.02 in.)
pore size
25-30 μm
basis weight
123 g/m2
capacity
high loading
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Grade 230 is a strong, thick creped paper widely used in oil chemistry. Retains coarse particles, has a fast filtration speed and high loading capacity.
Note; These papers should not be used for kjeldahl nitrogen analysis.
Features and Benefits
- Retains coarse particles
- Highspeedand loading capacity
- Strong, thick filter with crêped surface
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service