SYNA602424
Synthware™ modified anti-splash adapter
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A602424, Anti-splash adapter, splash head, bump trap
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
bottom joint: ST/NS24/40
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A602424
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
With two 4 mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service