Skip to Content
Merck

S135712

4,4'-(ETHANE-1,1-DIYL)DIPHENOL

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H14O2
CAS Number:
Molecular Weight:
214.266
MDL number:

Pricing and availability is not currently available.

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
62476L440873036
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

grade

for molecular biology

grade

for molecular biology

grade

-

grade

-

anion traces

sulfate (SO42-): ≤0.01%

anion traces

sulfate (SO42-): ≤50 mg/kg

anion traces

sulfate (SO42-): ≤0.01%

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ba: ≤0.003%, K: ≤0.01%, heavy metals (as Pb): ≤0.002%

cation traces

Al: ≤5 mg/kg, As: ≤0.2 mg/kg, Ba: ≤10 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤10 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

cation traces

Al: ≤0.0005%, Ba: ≤0.002%, Ca: ≤0.005%, Cu: ≤0.0005%, Fe: ≤0.002%, K: ≤0.005%, Mg: ≤0.005%, NH4+: ≤0.1%, Na: ≤0.005%, Pb: ≤0.001%, Zn: ≤0.001%

cation traces

Ba: ≤10 mg/kg, Ca: ≤20 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤10 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg

concentration

8 M

concentration

-

concentration

-

concentration

-

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service