Key Documents
BR474702
BRAND® microscope slide
cut edges, twin frosted end
Synonym(s):
Microscope slides, Microscopy
About This Item
UNSPSC Code:
41122600
NACRES:
MA.01
Pricing and availability is not currently available.
material
soda-lime glass
feature
cut edges, twin frosted end
manufacturer/tradename
BRAND 474702
size
76 mm × 26 mm × 1 mm
General description
  • Made from clear soda-lime glass, hydrolytic class 3.
  • Ground edges are recommended to reduce the risk of injury.
  • Thickness approx.: 1 mm
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)