Skip to Content
Merck
All Photos(3)

Key Documents

636959

Sigma-Aldrich

Titanium carbonitride

nanopowder, <150 nm particle size (spherical), ≥97% trace metals basis

Synonym(s):

Titanium carbide nitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ti2CN
CAS Number:
Molecular Weight:
121.75
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

Assay

≥97% trace metals basis

form

nanopowder

surface area

15-25 m2/g

particle size

<150 nm (spherical)

mp

>350 °C (lit.)

density

5.08 g/mL at 25 °C (lit.)

bulk density

0.23 g/mL

SMILES string

[Ti][Ti]C#N

InChI key

GMHFHWHEKOWDDD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service