Skip to Content
Merck
All Photos(1)

Documents

591629

Sigma-Aldrich

Phenol-18O

95 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H518OH
CAS Number:
Molecular Weight:
96.11
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

95 atom % 18O

form

solid

bp

182 °C (lit.)

mp

40-42 °C (lit.)

density

1.093 g/mL at 25 °C

mass shift

M+2

SMILES string

[18OH]c1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H/i7+2

InChI key

ISWSIDIOOBJBQZ-WGGUOBTBSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service