Skip to Content
Merck
All Photos(1)

Documents

282707

Sigma-Aldrich

Chlorotributylsilane

97%

Synonym(s):

Tributylchlorosilane, Chlorotributylsilane, Tributylsilyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3SiCl
CAS Number:
Molecular Weight:
234.88
Beilstein:
1742537
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

250-252 °C (lit.)

density

0.879 g/mL at 25 °C (lit.)

SMILES string

CCCC[Si](Cl)(CCCC)CCCC

InChI

1S/C12H27ClSi/c1-4-7-10-14(13,11-8-5-2)12-9-6-3/h4-12H2,1-3H3

InChI key

JSQJUDVTRRCSRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abishek Muralidhar et al.
Plant, cell & environment, 38(8), 1514-1527 (2014-12-30)
Aquatic organisms are often exposed to dramatic changes in salinity in the environment. Despite decades of research, many questions related to molecular and physiological mechanisms mediating sensing and adaptation to salinity stress remain unanswered. Here, responses of Vaucheria erythrospora, a
Branka D Živanović et al.
Planta, 242(4), 813-827 (2015-05-15)
Blue light signalling pathway in broad bean leaf epidermal cells includes key membrane transporters: plasma- and endomembrane channels and pumps of H (+) , Ca (2+) and K (+) ions, and plasma membrane redox system. Blue light signalling pathway in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service