Merck
  • Synthesis, structure, fullerene-binding and resolution of C3-symmetric cavitands with rigid and deep cavities.
Synthesis, structure, fullerene-binding and resolution of C3-symmetric cavitands with rigid and deep cavities.
Organic & biomolecular chemistry (2011-12-20)
Jin-Tao Yu, Zhi-Tang Huang, Qi-Yu Zheng
ABSTRACT
An efficient palladium-catalyzed Suzuki-Miyaura coupling method involving the reaction between CTV-Br(3) and a variety of aryl and heteroaryl boronic acids in the presence of indolyl phosphane ligands has been developed. This reaction procedure provided a series of C(3)-symmetric aryl-extended rigid cavitands for the first time. X-ray crystal structure analysis revealed that the phenyl substituted cavitand 5a has much larger rim edges and cavity height. This macrocyclic host adopts a linear head-to-tail "hand-shake" self-inclusion arrangement in the crystalline state. The fluorescence of 5a was considerably quenched upon the addition of C(60), with a binding constant of 78,700 ± 2300 dm(3) mol(-1) and a 1:1 stoichiometry according to the Job's plot. The interaction of C(60) with 5a in the excited state is stronger than that with CTV, which could be attributed to more binding sites in the extended arms of 5a. Moreover, optically active C(3)-symmetric cavitands (+)- and (-)-6 were easily obtained with high efficiency through chemical resolution.
Sigma-Aldrich
4-Pyridinylboronic acid, 90%