Skip to Content
Merck
All Photos(1)

Key Documents

814466

TSKgel® DEAE-5PW (13 µm) HPLC Columns

L x I.D. 2 cm × 20 mm, DEAE (diethylaminoethyl) phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
L × I.D.:
2 cm × 20 mm
particle size:
13 μm
matrix active group:
DEAE (diethylaminoethyl) phase
matrix:
hydroxylated methacrylic polymer

Product Name

TSKgel® DEAE-5PW Guards and Guardgel Kits, phase DEAE (diethylaminoethyl), L × I.D. 2 cm × 20 mm, 13 μm particle size, glass

material

glass

product line

TSKgel®

feature

counter ion Cl-

manufacturer/tradename

Tosoh 14466

parameter

≤1 M salt concentration
≤20% organic solvent
10-45 °C temperature
150 bar max. pressure
8 mL/min flow rate

technique(s)

LPLC: suitable

L × I.D.

2 cm × 20 mm

matrix

hydroxylated methacrylic polymer

matrix active group

DEAE (diethylaminoethyl) phase

particle size

13 μm

operating pH

2-12

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel® DEAE-5PW columns and guard columns are packed with porous hydrophilic polymer particles which are modified with a weak anion exchanger. They are used for the separation and analysis of proteins, nucleotides, nucleosides and other biologically active molecules. Columns are available in internal diameters varying from 2mm to 55mm ID. Column housings are either glass or stainless steel. They have the same backbone chemistry and selectivity as the bulk process scale resins Toyopearl® DEAE-650 and TSKgel DEAE-5PW (20) and (30).
TSKgel Guard cartridges require a holder, which is sold separately. For 2 mm cartridges, the holder is Part No. 819308. For the 3.2 mm cartridges used to protect 4.6 mm columns, the holder is Part No. 819018.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation
Toyopearl is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service