Skip to Content
Merck
All Photos(1)

Key Documents

52659-U

Supelco

Discovery® DSC-Si SPE Tube

bed wt. 10 g, volume 60 mL, pk of 16

Synonym(s):

Silica SPE cartridge tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

PE frit (20 μm porosity)
polypropylene hardware

Quality Level

composition

bed wt., 10 g

packaging

pk of 16

technique(s)

solid phase extraction (SPE): suitable

surface area

480 m2/g

volume

60 mL

matrix

silica gel base material (irregularly shaped, acid washed)

matrix active group

silica bonding

particle size

50 μm

pore size

70 Å pore size

application(s)

food and beverages

separation technique

normal phase

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Normal phase
Sample Matrix Compatibility: Organic solvents, oils, and lipids

  • Unbonded acid washed silica sorbent ideal for normal phase SPE and other modified flash techniques
  • Often used to separate or remove structurally similar molecules through successive elutions with increasingly polar solutions
  • The most polar normal phase sorbent available
  • Excellent capacity for purifying solution phase combinatorial chemistry reactions when removing target molecules from reaction by-products and excess reagents

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service