29603-U
Hamilton® syringe for CTC GC PAL
701N, volume 10 μL, needle size 23s ga, needle L 51 mm (2 in.)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
Recommended Products
material
cone tip stainless steel needle
needle size
23s ga
feature
needle type N (cemented needle)
point style 1
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 203361
needle L
51 mm (2 in.)
volume
10 μL
compatibility
for use with CTC GC PAL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Hamilton® syringe for CTC GC PAL offers the following advantages:
- Zero carry over.
- An innovative, cemented needle design that eliminates all interaction between the sample and the glued surface.
- Adjustable plunger protects the plunger tip from squeezing.
- Unique flange design that aligns the syringe in the proper position every time.
- More chemically resistant and temperature tolerant PTFE/polymer sealant for the plunger tip.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service